R&D Systems
Manufacturer Code:MAB39951SP
Catalog # MAB39951SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 925805R |
Immunogen | <em>E. coli</em>-derived recombinant human PARK-7 Ala2-Asp189 Accession # Q99497 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DJ-1; DJ1 DJ-1 DJ1FLJ34360 EC 3.4 FLJ27376 FLJ92274 Oncogene DJ1 Parkinson disease (autosomal recessive early onset) 7 Parkinson disease protein 7 protein DJ-1; epididymis secretory sperm binding protein Li 67p; Maillard deglycase; Oncogene DJ1; Parkinson disease (autosomal recessive, early onset) 7; Parkinson disease protein 7; parkinson protein 7; Parkinsonism-associated deglycase; Protein DJ-1; Protein/nucleic acid deglycase DJ-1
Gene Aliases: DJ-1; DJ1; HEL-S-67p; PARK7
UniProt ID: (Human) Q99497
Entrez Gene ID: (Human) 11315
Molecular Function:
RNA binding protein
cysteine protease
hydrolase
nucleic acid binding
protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.