Novus Biologicals
Manufacturer Code:NB1001194
Catalog # NB1001194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-AAQVKAPLVLKD corresponding to C-Terminus according to NP_009193.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: DJ-1; DJ-1 DJ1FLJ34360 EC 3.4 FLJ27376 FLJ92274 Oncogene DJ1 Parkinson disease (autosomal recessive early onset) 7 Parkinson disease protein 7 protein DJ-1; epididymis secretory sperm binding protein Li 67p; Maillard deglycase; Oncogene DJ1; Parkinson disease (autosomal recessive, early onset) 7; Parkinson disease protein 7; parkinson protein 7; Parkinsonism-associated deglycase; Protein DJ-1; Protein/nucleic acid deglycase DJ-1
Gene Aliases: DJ-1; DJ1; HEL-S-67p; PARK7
UniProt ID: (Human) Q99497
Entrez Gene ID: (Human) 11315
Molecular Function: RNA binding protein cysteine protease hydrolase nucleic acid binding protease transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.