Abnova
Manufacturer Code:MAB18383
Catalog # 89-338-308
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S121A-31 |
Immunogen | Recombinant protein corresponding to amino acids 11-664 (all but first 10 amino acids encompassing extracellular and cytoplasmic domains) of rat Panx2. |
Conjugate | ATTO488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: -; Pannexin-2
Gene Aliases: Panx2; Px2
UniProt ID: (Rat) P60571
Entrez Gene ID: (Rat) 362979
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.