Invitrogen
Manufacturer Code:PA542246
Catalog # PIPA542246
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human PANX3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gap junction protein pannexin 3; Pannexin-3; PANX3 PX3
Gene Aliases: 3230401P04; 4833413G11Rik; PANX3; PX3; si:dkey-263o8.2
UniProt ID: (Human) Q96QZ0, (Mouse) Q8CEG0, (Rat) P60572
Entrez Gene ID: (Dog) 609899, (Rabbit) 100350882, (Human) 116337, (Bovine) 538977, (Horse) 100064156, (Guinea pig) 100723554, (Mouse) 208098, (Rat) 315567, (Zebrafish) 100149056
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.