Novus Biologicals
Manufacturer Code:NBP191566
Catalog # NBP191566
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human PANX3. Peptide sequence IISELDKSYNRSIRLVQHMLKIRQKSSDPYVFWNELEKARKERYFEFPLL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gap junction protein pannexin 3; gap junction protein pannexin 3 pannexin 3 pannexin-3 PX3; Pannexin-3
Gene Aliases: PANX3; PX3
UniProt ID: (Human) Q96QZ0
Entrez Gene ID: (Human) 116337
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.