R&D Systems
Manufacturer Code:MAB7088SP
Catalog # MAB7088SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 719406 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Pannexin-2 Arg316-Lys456 Accession # Q96RD6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hPANX2 MGC119432 pannexin 2 pannexin-2 Pannexin2 PANX2 PX2; Pannexin-2
Gene Aliases: hPANX2; PANX2; PX2
UniProt ID: (Human) Q96RD6
Entrez Gene ID: (Human) 56666
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.