Invitrogen
Manufacturer Code:PA542245
Catalog # PIPA542245
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PANX2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Pannexin-2; PANX2 hPANX2 PX2
Gene Aliases: hPANX2; PANX2; PX2; si:ch211-192n14.2
UniProt ID: (Human) Q96RD6, (Mouse) Q6IMP4, (Rat) P60571
Entrez Gene ID: (Human) 56666, (Bovine) 539070, (Horse) 100056108, (Dog) 481195, (Mouse) 406218, (Guinea pig) 100726058, (Rat) 362979, (Zebrafish) 557828
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.