Novus Biologicals
Manufacturer Code:NBP159671
Catalog # NBP159671
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to PANX2(pannexin 2) The peptide sequence was selected from the N terminal of PANX2. Peptide sequence GTVLVPILLVTLVFTKNFAEEPIYCYTPHNFTRDQALYARGYCWTELRDA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPANX2 MGC119432 pannexin 2 pannexin-2 PX2; Pannexin-2
Gene Aliases: hPANX2; PANX2; PX2
UniProt ID: (Human) Q96RD6
Entrez Gene ID: (Human) 56666
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.