Invitrogen
Manufacturer Code:422800
Catalog # 42-280-0
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from the C-terminal region of mouse and rat pannexin 2 which differs by one non-conservative amino acid replacement from human and from predicted monkey bovine and dog sequences |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Pannexin-2; PANX2
Gene Aliases: hPANX2; PANX2; PX2
UniProt ID: (Human) Q96RD6, (Mouse) Q6IMP4, (Rat) P60571
Entrez Gene ID: (Human) 56666, (Mouse) 406218, (Rat) 362979
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.