R&D Systems
Manufacturer Code:MAB7097
Catalog # MAB7097
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 720505 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Pannexin-1 Pro298-Asp376 Accession # Q96RD7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: innexin; Innexin MGC21309 MRS1 MRS1innexin pannexin 1 pannexin-1 Pannexin1 PANX1 PX1 UNQ2529; Pannexin-1
Gene Aliases: MRS1; PANX1; PX1; UNQ2529; UNQ2529/PRO6028
UniProt ID: (Human) Q96RD7
Entrez Gene ID: (Human) 24145
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.