Invitrogen
Manufacturer Code:488000
Catalog # 48-800-0
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from the internal region of the mouse Pannexin 1 protein (AccessionNo. NP&lowbar062355) which is identical to rat and human sequence. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Pannexin-1; PANX1 Pannexin 1 Pannexin-1 PX1 MRS1 UNQ2529 UNQ2529/PRO6028
Gene Aliases: AI847747; Panx1; Px1
UniProt ID: (Rat) P60570, (Mouse) Q9JIP4
Entrez Gene ID: (Rat) 315435, (Mouse) 55991
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.