Novus Biologicals
Manufacturer Code:NBP273240C
Catalog # NBP273240C
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4B5 |
Immunogen | Full length recombinant protein of human Pancreatic Amylase Beta (NP_066188) produced in HEK293T cells. |
Conjugate | DyLight 650 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase 2B; Alpha-amylase 2B; alpha-amylase 2B AMY2 AMY2B amylase alpha 2B (pancreatic); Carcinoid alpha-amylase; glycogenase
Gene Aliases: AMY2; AMY2B; AMY3; HXA
UniProt ID: (Human) P19961
Entrez Gene ID: (Human) 280
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.