Invitrogen
Manufacturer Code:PA525330
Catalog # PIPA525330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 484-511 amino acids from the C-terminal region of human AMY2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase; AMY2 AMY2B PA 14-alpha-D-glucan glucanohydrolase alpha-amylase amylase pancreatic alpha-2A found in the pancreas glycogenase pancreatic alpha-amylase pancreatic amylase 2A pancreatic amylase alpha 2A; amylase 2a4; amylase 2a4, pancreatic; glycogenase; PA; Pancreatic alpha-amylase; pancreatic amylase alpha 2A
Gene Aliases: AMY2; Amy2-1; AMY2A; ENSMUSG00000074267; mAmy2-3; mAmy2-6; OTTMUSG00000022432; OTTMUSG00000022433; PA
UniProt ID: (Human) Q9UBH3, (Mouse) P00688
Entrez Gene ID: (Human) 279, (Mouse) 100043207, (Mouse) 100043684
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.