Invitrogen
Manufacturer Code:PA5117115
Catalog # PIPA5117115
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 242-511 of human AMY2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; 14-alpha-D-glucan glucanohydrolase 14-alpha-D-glucan glucanohydrolase 2B alpha-amylase Alpha-amylase 2B AMY2 Amy2-1 Amy2-2 Amy2a Amy2a1 Amy2a4 Amy2b AMY3 amylase amylase 2-2 pancreatic amylase 2a1 amylase 2a4 amylase 2a4 pancreatic amylase 2b amylase 2b pancreatic amylase alpha 2A (pancreatic) amylase alpha 2B (pancreatic) Amy-X carcinoid alpha-amylase ENSMUSG00000074267 glycogenase HXA mAmy2-1 mAmy2-3 mAmy2-6 OTTMUSG00000022432 OTTMUSG00000022433 OTTMUSG00000022462 PA Pancreatic alpha-amylase pancreatic amylase alpha 2A; Alpha-amylase 2B; amylase 2-2, pancreatic; amylase 2a4; amylase 2a4, pancreatic; amylase 2b; amylase 2b, pancreatic; Carcinoid alpha-amylase; glycogenase; PA; Pancreatic alpha-amylase; pancreatic amylase alpha 2A
Gene Aliases: Amy-X; AMY2; Amy2-1; Amy2-2; AMY2A; AMY2B; AMY3; ENSMUSG00000074267; HXA; mAmy2-1; mAmy2-3; mAmy2-6; OTTMUSG00000022432; OTTMUSG00000022433; OTTMUSG00000022462; PA
UniProt ID: (Human) Q9UBH3, (Human) P19961, (Mouse) P00688
Entrez Gene ID: (Human) 279, (Human) 280, (Mouse) 100043207, (Mouse) 100043684, (Mouse) 545562
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.