Invitrogen
Manufacturer Code:720260
Catalog # 72-026-0
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptides corresponding to human alpha-Amylase-2 (aa255-272 497-511 81-94) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase; 1,4-alpha-D-glucan glucanohydrolase 2B; 14-alpha-D-glucan glucanohydrolase 14-alpha-D-glucan glucanohydrolase 2B alpha-amylase Alpha-amylase 2B AMY2 Amy2-1 Amy2-2 Amy2a Amy2a1 Amy2a4 Amy2b AMY3 amylase amylase 2-2 pancreatic amylase 2a1 amylase 2a4 amylase 2a4 pancreatic amylase 2b amylase 2b pancreatic amylase alpha 2A (pancreatic) amylase alpha 2B (pancreatic) Amy-X carcinoid alpha-amylase ENSMUSG00000074267 glycogenase HXA mAmy2-1 mAmy2-3 mAmy2-6 OTTMUSG00000022432 OTTMUSG00000022433 OTTMUSG00000022462 PA Pancreatic alpha-amylase pancreatic amylase alpha 2A; Alpha-amylase 2B; Carcinoid alpha-amylase; glycogenase; PA; Pancreatic alpha-amylase; pancreatic amylase alpha 2A
Gene Aliases: AMY2; AMY2A; AMY2B; AMY3; HXA; PA
UniProt ID: (Human) Q9UBH3, (Human) P19961
Entrez Gene ID: (Human) 279, (Human) 280
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.