Novus Biologicals
Manufacturer Code:NBP273239H
Catalog # NBP273239H
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E3 |
Immunogen | Recombinant protein fragment corresponding to amino acids 135-408 of human Pancreatic Amylase Alpha (NP_000690) produced in E.coli. |
Conjugate | HRP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 1,4-alpha-D-glucan glucanohydrolase; AMY2 AMY2A amylase alpha 2A (pancreatic) AMY2B PA pancreatic alpha-amylase; glycogenase; PA; Pancreatic alpha-amylase; pancreatic amylase alpha 2A
Gene Aliases: AMY2; AMY2A; PA
UniProt ID: (Human) Q9UBH3
Entrez Gene ID: (Human) 279
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.