Proteintech
Manufacturer Code:213831AP150UL
Catalog # 50-173-3397
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Pan-PAX Fusion Protein Ag16018 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aniridia type II protein; B-cell lineage specific activator; B-cell-specific transcription factor; BSAP; Dickie's small eye; homeodomain protein PAX3; HuP1; HuP2; HuP48; Oculorhombin; optic disc coloboma; paired box gene 1; paired box gene 2; paired box gene 3; paired box gene 4; paired box gene 5; paired box gene 6; paired box gene 7; paired box gene 8; paired box gene 9; Paired box homeotic gene 1; paired box homeotic gene 2; paired box homeotic gene 3; paired box homeotic gene 5; paired box homeotic gene 6; paired box homeotic gene 7; paired box homeotic gene-6; paired box paired box 8 Paired box protein Pax Paired box protein Pax 8 Pan-PAX PAX 8 PAX8; Paired box protein Pax-1; Paired box protein Pax-2; Paired box protein Pax-3; Paired box protein Pax-4; Paired box protein Pax-5; Paired box protein Pax-6; Paired box protein Pax-7; Paired box protein Pax-8; Paired box protein Pax-9; paired box transcription factor PAX7; paired domain gene 3; paired domain gene 4; paired domain gene 5; paired domain gene 7; paired domain gene 8; paired domain gene 9; paired domain gene HuP2; paired domain gene HuP48; Pax-8 protein; PAX3/FKHR fusion; Pax5 variant 1; Pax5 variant 2; PAX7 transcriptional factor; pax9 {ECO:0000312|EMBL:BAE79272.1}; small eye; transcription factor PAX 5; wavy tail
Gene Aliases: 1500038E17Rik; AEY11; ALL3; AN; AN2; BSAP; CDHS; D11S812E; Dey; EBB-1; FSGS7; FVH1; Gsfaey11; hbs; hunchback; HUP1; HUP2; HUP48; KLP; KPD; MGDA; MODY9; OFC2; Opdc; PAPRS; Pax-1; Pax-2; Pax-3; Pax-4; Pax-5; Pax-6; Pax-7; Pax-8; Pax-9; PAX1; PAX2; PAX3; Pax4; PAX5; PAX6; PAX7; PAX7B; PAX8; PAX9; RGD1564360; RGD1565236; RMS2; Sey; Sp; splotch; STHAG3; un; undulated; WAGR; WS1; WS3; wt
UniProt ID: (Human) P15863, (Human) Q02962, (Human) P23760, (Human) Q02548, (Human) P26367, (Human) P23759, (Human) Q06710, (Human) P55771, (Mouse) P09084, (Mouse) P32114, (Mouse) P24610, (Rat) O88436, (Mouse) P32115, (Mouse) Q02650, (Mouse) P63015, (Rat) P63016, (Mouse) P47239, (Mouse) Q00288, (Rat) P51974, (Rat) Q2L4T2, (Mouse) P47242
Entrez Gene ID: (Human) 5075, (Human) 5076, (Human) 5077, (Human) 5078, (Human) 5079, (Human) 5080, (Human) 5081, (Human) 7849, (Human) 5083, (Rat) 311505, (Mouse) 18503, (Rat) 293992, (Mouse) 18504, (Rat) 114502, (Mouse) 18505, (Rat) 83630, (Mouse) 18506, (Rat) 500453, (Mouse) 18507, (Mouse) 18508, (Rat) 25509, (Rat) 500574, (Mouse) 18509, (Mouse) 18510, (Rat) 81819, (Rat) 362741, (Mouse) 18511
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.