Invitrogen
Manufacturer Code:41900380
Catalog # 50-112-3640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AE1/AE3 |
Conjugate | eFluor 570 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 40-kDa keratin intermediate filament; 47 kDa cytokeratin; 56 kDa cytokeratin; 57kd keratin; 57kDa keratin; 58 kDa cytokeratin; 60-kDa keratin; 65 kDa cytokeratin; 67 kDa cytokeratin; Cell proliferation-inducing gene 46 protein; CK-1; CK-10; CK-13; CK-14; CK-15; CK-16; CK-18; CK-19; CK-2e; CK-3; CK-4; CK-5; CK-6A; CK-6D; CK-7; CK-8; CK5 DDD EBS2 K5 KRT5A; cytokeratin 1; cytokeratin 10; cytokeratin 13; cytokeratin 14; cytokeratin 15; cytokeratin 16; cytokeratin 18; cytokeratin 19; cytokeratin 3; cytokeratin 4; cytokeratin 5; cytokeratin 6A; cytokeratin 6C; cytokeratin 6D; Cytokeratin 7; cytokeratin 8; Cytokeratin endo A; Cytokeratin endo B; Cytokeratin-1; Cytokeratin-10; Cytokeratin-13; Cytokeratin-14; Cytokeratin-15; Cytokeratin-16; Cytokeratin-18; Cytokeratin-19; Cytokeratin-2e; Cytokeratin-3; Cytokeratin-4; Cytokeratin-5; Cytokeratin-6A; cytokeratin-6C; Cytokeratin-6D; cytokeratin-6E; Cytokeratin-7; Cytokeratin-8; Cytoskeletal 57 kDa keratin; Endo B; epidermal keratin 10; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; epidermolysis bullosa simplex, Dowling-Meara, Koebner; epidermolytic hyperkeratosis 1; epithelial keratin 1; epithelial keratin 10; epithelial keratin 2e; Epithelial keratin-1; Epithelial keratin-2e; focal non-epidermolytic palmoplantar keratoderma; Hair alpha protein; Hom s 5; intermediate filament protein; K1; K10; K13; K14; K15; K16; K18; K19; K2e; K3; K3 keratin; K4; K5; K6A; K7; K8; kamp-keratin derived antimicrobial peptide; KDAMP; keratin 1 (epidermolytic hyperkeratosis); keratin 1, type II; keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris); keratin 10, type I; keratin 13, type I; keratin 14, type I; keratin 15, type I; keratin 16 (focal non-epidermolytic palmoplantar keratoderma); keratin 16, type I; keratin 18, type I; keratin 19, type I; keratin 2 epidermis; keratin 2, type II; keratin 2A (epidermal ichthyosis bullosa of Siemens); keratin 3, type II; keratin 4, type II; keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 6A, , type II; keratin 6A, type II; keratin 7, type II; keratin 8, type II; keratin 86; keratin complex 1, acidic, gene 10; keratin complex 1, acidic, gene 13; keratin complex 1, acidic, gene 14; keratin complex 1, acidic, gene 15; keratin complex 1, acidic, gene 16; keratin complex 1, acidic, gene 18; keratin complex 1, acidic, gene 19; keratin complex 2, basic, gene 1; keratin complex 2, basic, gene 17; keratin complex 2, basic, gene 4; keratin complex 2, basic, gene 5; keratin complex 2, basic, gene 6a; keratin complex 2, basic, gene 6c; keratin complex 2, basic, gene 7; keratin complex 2, basic, gene 8; keratin complex 2, gene 17; keratin complex 2, gene 6a; Keratin D; keratin intermediate filament 16a; keratin intermediate filament 16b; keratin K3; keratin K6h; keratin, 55K type II cytoskeletal; keratin, epidermal type II, K6A; keratin, simple epithelial type I, K7; Keratin, type I cytoskeletal 10; Keratin, type I cytoskeletal 13; Keratin, type I cytoskeletal 14; Keratin, type I cytoskeletal 15; Keratin, type I cytoskeletal 16; Keratin, type I cytoskeletal 18; Keratin, type I cytoskeletal 19; Keratin, type I cytoskeletal 59 kDa; keratin, type I, 40-kd; Keratin, type II cytoskeletal 1; Keratin, type II cytoskeletal 2 epidermal; Keratin, type II cytoskeletal 3; Keratin, type II cytoskeletal 4; Keratin, type II cytoskeletal 5; Keratin, type II cytoskeletal 6A; Keratin, type II cytoskeletal 7; Keratin, type II cytoskeletal 8; Keratin-1; Keratin-10; Keratin-13; Keratin-14; Keratin-15; keratin-15, basic; keratin-15, beta; Keratin-16; Keratin-18; Keratin-19; Keratin-2 epidermis; Keratin-2e; Keratin-3; Keratin-4; Keratin-5; Keratin-6-alpha; Keratin-6A; keratin-6C; Keratin-7; Keratin-8; krt2; krt5; krt7; mK6-alpha; Sarcolectin; suprabasal cytokeratin 10; tuftelin interacting protein 8; type I cytoskeletal 15; type II mesothelial keratin K7; Type-II keratin Kb1; type-II keratin Kb12; Type-II keratin Kb2; Type-II keratin Kb3; Type-II keratin Kb4; Type-II keratin Kb5; Type-II keratin Kb6; Type-II keratin Kb7; Type-II keratin Kb8
Gene Aliases: 2310016L08Rik; 3300001P10Rik; AA960620; AI324768; AI528832; AI626930; AI663979; AL022697; AU019895; AW108092; AW146334; BB005427; BCIE; BIE; CARD2; CK-10; CK-18; CK-2e; CK-3; CK-4; CK-6C; CK-6E; CK-8; CK1; CK10; CK13; CK14; CK15; CK16; CK18; CK19; CK3; CK4; CK5; CK6A; CK6C; CK6D; CK7; CK8; CYK18; CYK4; CYK8; D130054E02Rik; D15Wsu77e; DDD; DDD1; EBS2; EBS3; EBS4; EGK_03684; EGK_03685; EHK; EHK1; EndoA; EndoC; EPPK; FNEPPK; K1; K10; K13; K14; K15; K16; K18; K19; K1C1; K1CO; K1CP; K1CS; K2C7; K2C8; K2E; K3; K4; K5; K6A; K6C; K6D; K7; K8; KER1; Ker10; Ker2; Kerd; KO; KPP; Krt-1.10; Krt-1.13; Krt-1.14; Krt-1.19; Krt-2.1; Krt-2.4; Krt-2.8; KRT1; Krt1-10; Krt1-13; Krt1-14; Krt1-15; Krt1-16; Krt1-18; Krt1-19; KRT10; KRT13; KRT14; KRT15; KRT16; KRT16A; KRT18; KRT19; KRT1A; KRT2; Krt2-1; Krt2-17; Krt2-2; Krt2-4; Krt2-5; Krt2-6; Krt2-6a; Krt2-6c; Krt2-7; Krt2-8; KRT2A; KRT2E; KRT3; KRT4; KRT5; KRT5A; Krt6; KRT6A; KRT6C; KRT6D; KRT7; KRT8; Krt86; KRTA; KRTB; KRTE; mK6[a]; MK6a; NEPPK; NFJ; PC1; PC3; PIG46; SCL; Tfip8; WSN1; WSN2
UniProt ID: (Dog) Q6EIY9, (Human) P04264, (Human) P13645, (Human) P13646, (Human) P02533, (Human) P19012, (Human) P08779, (Human) P05783, (Human) P08727, (Human) P35908, (Human) P12035, (Rabbit) Q29426, (Human) P19013, (Human) P13647, (Human) P02538, (Human) P08729, (Human) P05787, (Mouse) P04104, (Mouse) Q9CXH6, (Mouse) P08730, (Mouse) Q61781, (Mouse) Q61414, (Mouse) Q9Z2K1, (Mouse) P05784, (Mouse) P19001, (Mouse) Q3TTY5, (Mouse) P07744, (Mouse) Q922U2, (Mouse) P50446, (Mouse) Q9DCV7, (Mouse) P11679
Entrez Gene ID: (Dog) 444857, (Rhesus monkey) 699320, (Rabbit) 100353415, (Human) 3848, (Rabbit) 100009029, (Dog) 491006, (Human) 3858, (Rhesus monkey) 702153, (Human) 3860, (Rhesus monkey) 706830, (Dog) 490983, (Rhesus monkey) 100426180, (Human) 3861, (Dog) 100685323, (Rabbit) 100341120, (Rhesus monkey) 106994041, (Dog) 100685404, (Human) 3866, (Dog) 490978, (Human) 3868, (Rhesus monkey) 707236, (Rabbit) 100341376, (Human) 3875, (Dog) 477601, (Rabbit) 100354174, (Human) 3880, (Dog) 100685481, (Rabbit) 100353162, (Rhesus monkey) 699567, (Human) 3849, (Dog) 444850, (Human) 3850, (Dog) 100683401, (Rabbit) 100353668, (Rhesus monkey) 100425096, (Human) 3851, (Dog) 100682832, (Rhesus monkey) 700060, (Rabbit) 100348388, (Human) 3852, (Rhesus monkey) 100424400, (Rabbit) 100352658, (Dog) 100687760, (Dog) 486523, (Human) 3853, (Dog) 477602, (Human) 3855, (Rabbit) 100345579, (Rhesus monkey) 697425, (Rabbit) 100348891, (Human) 3856, (Dog) 486513, (Rhesus monkey) 700449, (Mouse) 16678, (Mouse) 16661, (Mouse) 16663, (Mouse) 16664, (Mouse) 16665, (Mouse) 16666, (Mouse) 16668, (Mouse) 16669, (Mouse) 16681, (Mouse) 16682, (Mouse) 110308, (Mouse) 16687, (Mouse) 110310, (Mouse) 16691
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.