OriGene
Manufacturer Code:TA190311
Catalog # 50-167-1286
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5G2 |
Immunogen | Synthetic peptide corresponding to the conserved region of human cadherin family. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cadherin 1, E-cadherin (epithelial); cadherin 1, type 1; cadherin 1, type 1, E-cadherin (epithelial); cadherin 2, type 1, N-cadherin (neuronal); cadherin 3, type 1, P-cadherin (placental); cadherin 4, type 1, preproprotein; cadherin 4, type 1, R-cadherin (retinal); Cadherin-1; Cadherin-2; Cadherin-3; Cadherin-4; calcium-dependent adhesion protein, epithelial; calcium-dependent adhesion protein, neuronal; calcium-dependent adhesion protein, placental; CAM 120/80; CD324; CD325; CDw325; cell-CAM 120/80; E-Cad/CTF1; E-Cad/CTF2; E-Cad/CTF3; E-Cadherin; E-cadherin 1; Epithelial cadherin; N-cadherin; N-cadherin 1; Neural cadherin; P-cadherin; Pan-CDH; Placental cadherin; R-CAD; R-cadherin; Retinal cadherin; Uvomorulin
Gene Aliases: Arc-1; CAD4; CD324; CD325; CDH1; CDH2; CDH3; CDH4; CDHE; CDHN; CDHP; CDw325; ECAD; HJMD; LCAM; NCAD; PCAD; R-CAD; RCAD; UVO
UniProt ID: (Human) P12830, (Human) P19022, (Human) P22223, (Human) P55283
Entrez Gene ID: (Human) 999, (Human) 1000, (Human) 1001, (Human) 1002
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.