Invitrogen
Manufacturer Code:MA527722
Catalog # PIMA527722
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S388A-60 |
Immunogen | Fusion protein amino acids 1-1881 (full-length) of human Ankyrin-R |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ANK-1; ANK-2; ANK-3; ank2; ankyrin 1, erythrocytic; ankyrin 1, erythroid; ankyrin 2, brain; ankyrin 3 (G); ankyrin 3, epithelial; ankyrin B; ankyrin, brain; Ankyrin-1; Ankyrin-1 Erythrocyte Ankyrin ANK-1 ANK rCG&lowbar43073; Ankyrin-2; ankyrin-2, nonerythrocytic; Ankyrin-3; Ankyrin-B; Ankyrin-G; Ankyrin-R; Brain ankyrin; brain-specific ankyrin-G; Erythrocyte ankyrin; Non-erythroid ankyrin; normoblastic anemia
Gene Aliases: 2900054D09Rik; AI314020; AI835472; ANK; Ank-1; ANK-2; ANK-3; ANK1; ANK2; ANK3; ANKB; AnkG; Ankyrin-3; ANKYRIN-G; AW491075; brank-2; LQT4; MRT37; nb; pale; SPH1; SPH2
UniProt ID: (Human) P16157, (Human) Q01484, (Human) Q12955, (Mouse) Q02357, (Mouse) Q8C8R3, (Rat) O70511, (Mouse) G5E8K5
Entrez Gene ID: (Human) 286, (Human) 287, (Human) 288, (Mouse) 11733, (Rat) 306570, (Mouse) 109676, (Rat) 362036, (Rat) 361833, (Mouse) 11735
Molecular Function: cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.