Proteintech
Manufacturer Code:108531AP150UL
Catalog # 50-172-6657
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Palladin Fusion Protein Ag1288 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 90kDa actin-associated protein palladin; CGI 151 KIAA0992 palladin PALLD PNCA1 Sarcoma antigen NY SAR 77 SIH002; myoneurin; Palladin; palladin isoform; Sarcoma antigen NY-SAR-77; SIH002
Gene Aliases: 2410003B16Rik; 6030492A02; CGI-151; CGI151; KIAA0992; MYN; PALLD; PNCA1; SIH002
UniProt ID: (Human) Q8WX93, (Mouse) Q9ET54
Entrez Gene ID: (Human) 23022, (Mouse) 72333
Molecular Function:
cell adhesion molecule
cytokine receptor
defense/immunity protein
hydrolase
immunoglobulin receptor superfamily
immunoglobulin superfamily cell adhesion molecule
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.