Abnova
Manufacturer Code:PAB11726
Catalog # 89-115-332
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 525-545 of rat Pak1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rabbit polyclonal antibody raised against synthetic peptide of Pak1.
For Research Use Only
Protein Aliases: Alpha-PAK; p21 (CDKN1A)-activated kinase 1; p21-activated kinase 1; p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related); p68-PAK; PAK-1; Protein kinase MUK2; Serine/threonine-protein kinase PAK 1
Gene Aliases: Pak1
UniProt ID: (Rat) P35465
Entrez Gene ID: (Rat) 29431
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.