Invitrogen
Manufacturer Code:OSP00020W
Catalog # PIOSP00020W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Fruit fly |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from 1t cytoplasmic loop of drosophila PYX. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AT05393p; AT29936p; CG17142-PA; CG17142-PB; pyx-PA; pyx-PB; Transient receptor potential channel pyrexia
Gene Aliases: CG17142; CT33412; Dmel\CG17142; Dmel_CG17142; dTRPA2; Pyrexia; Pyx; trpA2
UniProt ID: (Fruit fly) Q9W0T5
Entrez Gene ID: (Fruit fly) 38037
Molecular Function:
ion channel
transporter
voltage-gated ion channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.