R&D Systems
Manufacturer Code:MAB6210
Catalog # MAB6210
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 592918 |
Immunogen | Phosphopeptide containing the human PYK2/FAK2 Y402 site Accession # Q14289 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CADTK; CADTK CADTKCAK-beta CAK beta CAKB Calcium-dependent tyrosine kinase Cell adhesion kinase beta EC 2.7.10 FADK2 FAK2 FAK2EC 2.7.10.2 Focal adhesion kinase 2 FRNK PKB Proline-rich tyrosine kinase 2 protein kinase B protein tyrosine kinase 2 beta protein-tyrosine kinase 2-beta PTK PTK2B PTK2B protein tyrosine kinase 2 beta PYK2Related adhesion focal tyrosine kinase RAFTK RAFTKFADK 2; CAK-beta; Calcium-dependent tyrosine kinase; Calcium-regulated non-receptor proline-rich tyrosine kinase; Cell adhesion kinase beta; FADK 2; Focal adhesion kinase 2; Proline-rich tyrosine kinase 2; protein kinase B; Protein-tyrosine kinase 2-beta; PTK2B protein tyrosine kinase 2 beta; RAFTK; Related adhesion focal tyrosine kinase
Gene Aliases: CADTK; CAKB; FADK2; FAK2; PKB; PTK; PTK2B; PYK2; RAFTK
UniProt ID: (Human) Q14289
Entrez Gene ID: (Human) 2185
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.