Invitrogen
Manufacturer Code:PA5110248
Catalog # PIPA5110248
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human PYGL (NP_002854.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycogen phosphorylase liver form GPLL GSD6 Lgp PYGL; Glycogen phosphorylase, liver form; liver glycogen phosphorylase
Gene Aliases: GSD6; Lgp; PYGL
UniProt ID: (Human) P06737, (Mouse) Q9ET01, (Rat) P09811
Entrez Gene ID: (Human) 5836, (Mouse) 110095, (Rat) 64035
Molecular Function:
phosphorylase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.