Abnova
Manufacturer Code:H00029108B02
Catalog # 89-007-491
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | PYCARD (NP_037390 1 a.a. ~ 195 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human PYCARD protein.
For Research Use Only
Protein Aliases: Apoptosis-associated speck-like protein containing a CARD; ASC; CARD5; Caspase recruitment domain-containing protein 5; hASC; MGC10332; PYD and CARD domain-containing protein; Target of methylation-induced silencing 1; TMS; TMS-1; TMS1
Gene Aliases: ASC; CARD5; PYCARD; TMS; TMS-1; TMS1
UniProt ID: (Human) Q9ULZ3
Entrez Gene ID: (Human) 29108
Molecular Function:
cysteine protease
enzyme modulator
hydrolase
protease
protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.