Novus Biologicals
Manufacturer Code:NBP273752PE
Catalog # NBP273752PE
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B12 |
Immunogen | Full length recombinant protein of human PYCRL (NP_075566) produced in HEK293T cells. |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 1.5.1.2 FLJ13852 P5C reductase 3 P5CR 3 pyrroline-5-carboxylate reductase 3 pyrroline-5-carboxylate reductase-like Pyrroline-5-carboxylate reductase-like protein; P5C reductase 3; P5CR 3; Pyrroline-5-carboxylate reductase 3; Pyrroline-5-carboxylate reductase-like protein
Gene Aliases: PYCR3; PYCRL
UniProt ID: (Human) Q53H96
Entrez Gene ID: (Human) 65263
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.