Invitrogen
Manufacturer Code:PA530141
Catalog # PIPA530141
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 277 of Human PYCR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARCL2B ARCL3B P5C P5CR PIG45 PP222 PRO3 PYCR P5C reductase 1 P5CR 1 mitochondrial pyrroline-5-carboxylate reductase 1 proliferation-inducing protein 45 pyrroline-5-carboxylate reductase 1 mitochondrial; P5C reductase 1; proliferation-inducing protein 45; Pyrroline-5-carboxylate reductase 1, mitochondrial
Gene Aliases: ARCL2B; ARCL3B; P5C; P5CR; PIG45; PP222; PRO3; PYCR; PYCR1
UniProt ID: (Human) P32322
Entrez Gene ID: (Human) 5831
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.