Proteintech
Manufacturer Code:221501AP150UL
Catalog # 50-173-3745
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PYCR1 Fusion Protein Ag17745 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: P5C P5C reductase 1 P5CR P5CR 1 PIG45 PP222 PYCR PYCR1; P5C reductase 1; P5CR 1; proliferation-inducing protein 45; Pyrroline-5-carboxylate reductase 1, mitochondrial
Gene Aliases: ARCL2B; ARCL3B; P5C; P5CR; PIG45; PP222; PRO3; PYCR; PYCR1
UniProt ID: (Human) P32322, (Mouse) Q922W5
Entrez Gene ID: (Human) 5831, (Mouse) 209027, (Rat) 287877
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.