Novus Biologicals
Manufacturer Code:NBP250519AF488
Catalog # NBP250519AG
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | V11P4G11&asteriskE7 |
Immunogen | Ovalbumin-conjugated synthetic peptide EMRSMNEEYT (C-terminal sequence) |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Nuclear receptor subfamily 1 group I member 2; nuclear receptor subfamily 1 group I member 2 nuclear receptor subfamily 1 group I member 2 ONR1 Orphan nuclear receptor PAR1 Orphan nuclear receptor PXR PAR PAR1 PAR2 Pregnane X receptor PRR PXRPARq SAR Steroid and xenobiotic receptor SXRBXR; nuclear receptor subfamily 1, group I, member 2; Orphan nuclear receptor PAR1; Orphan nuclear receptor PXR; pregnane X nuclear receptor variant 2; Pregnane X receptor; Steroid and xenobiotic receptor; SXR
Gene Aliases: BXR; NR1I2; ONR1; PAR; PAR1; PAR2; PARq; PRR; PXR; SAR; SXR
UniProt ID: (Human) O75469
Entrez Gene ID: (Human) 8856
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.