Invitrogen
Manufacturer Code:PA572551
Catalog # PIPA572551
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 100-127 amino acids from the Central region of human NR1I2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Nuclear receptor subfamily 1 group I member 2; nuclear receptor subfamily 1, group I, member 2; Orphan nuclear receptor PAR1; Orphan nuclear receptor PXR; pregnane X nuclear receptor variant 2; Pregnane X receptor; PXR NR1I2 nuclear receptor subfamily 1 group I member 2 BXR ONR1 PAR PAR1 PAR2 PARq PRR SAR SXR orphan nuclear receptor prege X receptor steroid and xenobiotic receptor; Steroid and xenobiotic receptor; SXR
Gene Aliases: BXR; NR1I2; ONR1; PAR; PAR1; PAR2; PARq; PRR; PXR; SAR; SXR
UniProt ID: (Human) O75469
Entrez Gene ID: (Human) 8856
Molecular Function:
nuclear hormone receptor
nucleic acid binding
receptor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.