Invitrogen
Manufacturer Code:PA5116715
Catalog # PIPA5116715
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human PXR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BXR mPXR NR1I2 nuclear receptor subfamily 1 group I member 2 nuclear receptor subfamily 1 group 1 member 2 nuclear receptor subfamily 1 group I member 2 ONR 1 ONR1 Orphan nuclear receptor PAR1 orphan nuclear receptor PXR PAR PAR 1 PAR 2 PAR q PAR1 PAR2 PARq pregnane X nuclear receptor variant 2 Pregnane X receptor pregnane X receptor (nuclear receptor sub family 1 group I member 2) PRR PXR PXR.1 PXR.2 PXR1 SAR Steroid and xenobiotic receptor SXR; Nuclear receptor subfamily 1 group I member 2; nuclear receptor subfamily 1, group 1, member 2; nuclear receptor subfamily 1, group I, member 2; Orphan nuclear receptor PAR1; Orphan nuclear receptor PXR; pregnane X nuclear receptor variant 2; Pregnane X receptor; pregnane X receptor (nuclear receptor sub family 1, group I, member 2); Steroid and xenobiotic receptor; SXR
Gene Aliases: BXR; mPXR; NR1I2; ONR1; PAR; PAR1; PAR2; PARq; PRR; PXR; PXR.1; PXR.2; PXR1; SAR; SXR
UniProt ID: (Human) O75469, (Rat) Q9R1A7, (Mouse) O54915
Entrez Gene ID: (Human) 8856, (Rat) 84385, (Mouse) 18171
Molecular Function: nuclear hormone receptor nucleic acid binding receptor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.