Abnova
Manufacturer Code:PAB7904
Catalog # 89-133-484
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S178 of human PXN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PXN.
For Research Use Only
Protein Aliases: FLJ16691; Paxillin; testicular tissue protein Li 134
Gene Aliases: PXN
UniProt ID: (Human) P49023
Entrez Gene ID: (Human) 5829
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.