Novus Biologicals
Manufacturer Code:NBP29464320UL
Catalog # NB023811
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human PEX2 (NP_001165558.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 35 kDa peroxisomal membrane protein; PAF-1; PAF-1 PAF1peroxisomal membrane protein 3 (35kD Zellweger syndrome) Peroxin-2 peroxisomal biogenesis factor 235 kDa peroxisomal membrane protein Peroxisomal membrane protein 3 peroxisomal membrane protein 3 35kDa Peroxisome assembly factor 1 peroxisome biogenesis factor 2 PMP35PMP3 PXMP3peroxisome assembly factor-1 RNF72RING finger protein 72 ZWS3; Peroxin-2; Peroxisomal membrane protein 3; peroxisomal membrane protein 3, 35kDa; Peroxisome assembly factor 1; peroxisome assembly factor-1; Peroxisome biogenesis factor 2; RING finger protein 72
Gene Aliases: PAF1; PBD5A; PBD5B; PEX2; PMP3; PMP35; PXMP3; RNF72; ZWS3
UniProt ID: (Human) P28328
Entrez Gene ID: (Human) 5828
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.