Proteintech
Manufacturer Code:221631AP150UL
Catalog # 50-173-3752
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PXMP3 Fusion Protein Ag17815 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 35 kDa peroxisomal membrane protein; PAF 1 PAF1 Peroxin 2 Peroxisomal membrane protein 3 Peroxisome assembly factor 1 Peroxisome biogenesis factor 2 PEX2 PMP3 PMP35 PXMP3 RING finger protein 72 RNF72; PAF-1; Peroxin-2; Peroxisomal membrane protein 3; peroxisomal membrane protein 3, 35 kDa; peroxisomal membrane protein 3, 35kDa; Peroxisome assembly factor 1; peroxisome assembly factor-1; Peroxisome biogenesis factor 2; RING finger protein 72; Zellweger syndrome homolog
Gene Aliases: D3Ertd138e; PAF-1; PAF1; PBD5A; PBD5B; PEX2; PMP3; PMP35; PXMP3; RNF72; ZWS3
UniProt ID: (Human) P28328, (Mouse) P55098
Entrez Gene ID: (Human) 5828, (Mouse) 19302
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.