Novus Biologicals
Manufacturer Code:NBP259125
Catalog # NBP259125
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Maps to a region between 528 and 578 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ20335 K-ATPase K-ATPase long form Modulator of Na MONaKA PX domain containing serine/threonine kinase PX domain-containing protein kinase-like protein PX ser/thr kinase v2 PX serine/threonine kinase; Modulator of Na,K-ATPase; modulator of Na,K-ATPase long form; MONaKA; PX domain-containing protein kinase-like protein; PX ser/thr kinase v2; PX serine/threonine kinase; PXK
Gene Aliases: MONaKA; PXK
UniProt ID: (Human) Q7Z7A4
Entrez Gene ID: (Human) 54899
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.