Abnova
Manufacturer Code:H00027166A01
Catalog # 89-106-756
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 3 a.a. &bsim 100 a.a) partial recombinant protein with GST tag.,PX19 (NP_037369 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant PX19.
For Research Use Only
Protein Aliases: 25 kDa protein of relevant evolutionary and lymphoid interest; CGI-106; MGC87972; PRELI; PRELI domain-containing protein 1, mitochondrial; PX19; Px19-like protein
Gene Aliases: CGI-106; PRELI; PRELID1; PX19; SBBI12
UniProt ID: (Human) Q9Y255
Entrez Gene ID: (Human) 27166
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.