Novus Biologicals
Manufacturer Code:NBP273733FR
Catalog # NBP273733FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5F12 |
Immunogen | Full length recombinant protein of human PUS7 (NP_061915) produced in HEK293T cells. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 5.4.99.- FLJ20485 KIAA1897 MGC17720 pseudouridylate synthase 7 homolog pseudouridylate synthase 7 homolog (S. cerevisiae); Pseudouridylate synthase 7 homolog
Gene Aliases: KIAA1897; PUS7
UniProt ID: (Human) Q96PZ0
Entrez Gene ID: (Human) 54517
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.