Invitrogen
Manufacturer Code:PA524321
Catalog # PIPA524321
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 35-64 amino acids from the N-terminal region of human PUS3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610020J05Rik tRNA pseudouridine synthase 3 tRNA pseudouridine(38/39) synthase tRNA pseudouridylate synthase 3 tRNA-uridine isomerase 3; tRNA pseudouridine synthase 3; tRNA pseudouridine(38/39) synthase; tRNA pseudouridylate synthase 3; tRNA-uridine isomerase 3
Gene Aliases: 2610020J05Rik; FKSG32; PUS3
UniProt ID: (Human) Q9BZE2
Entrez Gene ID: (Human) 83480
Molecular Function: lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.