Proteintech
Manufacturer Code:172481AP150UL
Catalog # 50-173-1462
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PUS3 Fusion Protein Ag9983 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610020J05Rik FKSG32 pseudouridylate synthase 3 PUS3 tRNA pseudouridine synthase 3 tRNA uridine isomerase 3; pseudouridine synthase 3; tRNA pseudouridine synthase 3; tRNA pseudouridine(38/39) synthase; tRNA pseudouridylate synthase 3; tRNA-uridine isomerase 3
Gene Aliases: 2610020J05Rik; 5730412F04Rik; FKSG32; PUS3; RGD1310757
UniProt ID: (Human) Q9BZE2, (Mouse) Q9JI38
Entrez Gene ID: (Human) 83480, (Mouse) 67049, (Rat) 315554
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.