Invitrogen
Manufacturer Code:PA590861
Catalog # PIPA590861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 290-529 of human PUS10 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: coiled-coil domain containing 139; Coiled-coil domain-containing protein 139; Psi55 synthase; putative tRNA pseudouridine synthase Pus10; tRNA pseudouridine 55 synthase; tRNA pseudouridine synthase Pus10; tRNA pseudouridylate synthase; tRNA-uridine isomerase
Gene Aliases: 2810013G11Rik; 4933435A13Rik; AU014648; C77560; Ccdc139; Pus10; RGD1306402
UniProt ID: (Mouse) Q9D3U0
Entrez Gene ID: (Rat) 305583, (Mouse) 74467
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.