Invitrogen
Manufacturer Code:PA585285
Catalog # PIPA585285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human PUS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC11268 antibody MLASA1 antibody PUS1 antibody tRNA pseudouridine(38-40) synthase antibody tRNA uridine isomerase I antibody tRNA pseudouridylate synthase I antibody tRNA pseudouridine synthase A mitochondrial antibody tRNA-uridine isomerase I antibody mitochondrial tRNA pseudouridine synthase A antibody pseudouridylate synthase 1 antibody; mitochondrial tRNA pseudouridine synthase A; tRNA pseudouridine synthase A; tRNA pseudouridine synthase A, mitochondrial; tRNA pseudouridine(38-40) synthase; tRNA pseudouridylate synthase I; tRNA uridine isomerase I; tRNA-uridine isomerase I
Gene Aliases: A730013B20Rik; MLASA1; MNCb-0873; MPUS1; mPus1p; PP8985; PUS1
UniProt ID: (Human) Q9Y606, (Mouse) Q9WU56
Entrez Gene ID: (Human) 80324, (Mouse) 56361
Molecular Function: lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.