Invitrogen
Manufacturer Code:PA568838
Catalog # PIPA568838
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse PUS1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MLASA1 mitochondrial tRNA pseudouridine synthase A tRNA pseudouridine synthase A mitochondrial tRNA pseudouridine(38-40) synthase tRNA pseudouridylate synthase I tRNA uridine isomerase I tRNA-uridine isomerase I; pseudouridine synthase 1; pus1 {ECO:0000312|RGD:1311871}; tRNA pseudouridine synthase A; tRNA pseudouridine synthase A, mitochondrial; tRNA pseudouridine(38-40) synthase; tRNA pseudouridylate synthase I; tRNA-uridine isomerase I
Gene Aliases: A730013B20Rik; MNCb-0873; MPUS1; mPus1p; Pus1
UniProt ID: (Rat) Q4KM92, (Mouse) Q9WU56
Entrez Gene ID: (Rat) 304567, (Mouse) 56361
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.