Novus Biologicals
Manufacturer Code:NBP256899
Catalog # NBP256899
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LKVWLIDDILEKINSHLPSHIRILGLKRVTGGFNSKNRCDARTYCYLLPTFAFAHKDRDVQDETYRLSAETLQQVNRLLACYKGTHNFHNFTSQ |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 5.4.99.12 MGC11268 mitochondrial tRNA pseudouridine synthase A MLASA1 pseudouridylate synthase 1 tRNA pseudouridine synthase A mitochondrial tRNA pseudouridylate synthase I tRNA uridine isomerase I tRNA-uridine isomerase I; mitochondrial tRNA pseudouridine synthase A; tRNA pseudouridine synthase A; tRNA pseudouridine(38-40) synthase; tRNA pseudouridylate synthase I; tRNA uridine isomerase I; tRNA-uridine isomerase I
Gene Aliases: MLASA1; PP8985; PUS1
UniProt ID: (Human) Q9Y606
Entrez Gene ID: (Human) 80324
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.