Novus Biologicals
Manufacturer Code:NBP297161R
Catalog # NB101127
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human PUM2 fragment. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ36528 KIAA0235PUMH2PUML2 MGC138251 MGC138253 pumilio (Drosphila) homolog 2 pumilio homolog 2 pumilio homolog 2 (Drosophila) Pumilio-2; Pumilio homolog 2; pumilio RNA-binding family member 2; Pumilio-2
Gene Aliases: KIAA0235; PUM2; PUMH2; PUML2
UniProt ID: (Human) Q8TB72
Entrez Gene ID: (Human) 23369
Molecular Function: RNA binding protein mRNA processing factor nucleic acid binding translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.