Novus Biologicals
Manufacturer Code:NB100387
Catalog # NB100387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residues 100 and 150 of human Pumilio 2 using the numbering given in entry NP_056132.1 (GeneID 23369). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ36528 KIAA0235PUMH2PUML2 MGC138251 MGC138253 pumilio (Drosphila) homolog 2 pumilio homolog 2 pumilio homolog 2 (Drosophila) Pumilio-2; Pumilio homolog 2; pumilio RNA-binding family member 2; Pumilio-2
Gene Aliases: KIAA0235; PUM2; PUMH2; PUML2
UniProt ID: (Human) Q8TB72
Entrez Gene ID: (Human) 23369
Molecular Function: RNA binding protein mRNA processing factor nucleic acid binding translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.