Abnova
Manufacturer Code:PAB6759
Catalog # 89-132-663
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of human PUM2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of PUM2.
For Research Use Only
Protein Aliases: FLJ36528; KIAA0235; MGC138251; MGC138253; PUMH2; Pumilio homolog 2; pumilio RNA-binding family member 2; Pumilio-2; PUML2
Gene Aliases: KIAA0235; PUM2; PUMH2; PUML2
UniProt ID: (Human) Q8TB72
Entrez Gene ID: (Human) 23369
Molecular Function: RNA binding protein mRNA processing factor nucleic acid binding translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.