Proteintech
Manufacturer Code:115861AP150UL
Catalog # 50-172-7288
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PUM2 Fusion Protein Ag2179 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA0235 PUM2 PUMH2 Pumilio 2 Pumilio homolog 2 pumilio homolog 2 (Drosophila) PUML2; PUM1; pumilio 2; Pumilio homolog 2; pumilio RNA-binding family member 2; Pumilio-2
Gene Aliases: 5730503J23Rik; KIAA0235; mKIAA0235; PUM2; PUMH2; PUML2; Pumm2
UniProt ID: (Human) Q8TB72, (Mouse) Q80U58
Entrez Gene ID: (Human) 23369, (Rat) 298874, (Mouse) 80913
Molecular Function: RNA binding protein mRNA processing factor nucleic acid binding translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.