Novus Biologicals
Manufacturer Code:NBP275638
Catalog # NBP275638
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG37-75 |
Immunogen | Recombinant protein within human Pumilio 1 aa 1-200. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: HsPUM; HSPUM KIAA0099PUML1 PUMH PUMH1pumilio homolog 1 pumilio (Drosophila) homolog 1 pumilio homolog 1 (Drosophila) Pumilio-1; Pumilio homolog 1; pumilio RNA-binding family member 1; pumilio-1
Gene Aliases: HSPUM; KIAA0099; PUM1; PUMH; PUMH1; PUML1
UniProt ID: (Human) Q14671
Entrez Gene ID: (Human) 9698
Molecular Function:
RNA binding protein
mRNA processing factor
nucleic acid binding
translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.