R&D Systems
Manufacturer Code:MAB5870SP
Catalog # MAB5870SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 732322 |
Immunogen | <EM>E. coli</EM>-derived recombinant human PU.1/Spi-1 Met1-Lys169 Accession # NP_001074016 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 31 kDa transforming protein; 31 kDa-transforming protein; hematopoietic transcription factor PU.1; hematopoietic transcription factor PU.1 OF PU.1 SFPI1 SFPI131 kDa transforming protein SPI1 SPI-1 SPI-1 proto-oncogene SPI-A spleen focus forming virus (SFFV) proviral integration oncogene spi131 kDa-transforming protein transcription factor PU.1; spleen focus forming virus (SFFV) proviral integration oncogene spi1; Transcription factor PU.1
Gene Aliases: OF; PU.1; SFPI1; SPI-1; SPI-A; SPI1
UniProt ID: (Human) P17947
Entrez Gene ID: (Human) 6688
Molecular Function: nucleic acid binding signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.